Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl

Catalog No. AC196085000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196085000 500 mL
AC196081000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196085000 Supplier Thermo Scientific Chemicals Supplier No. 196085000
Add to Cart

CAS: 7439-93-2 | Li | 6.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Li in 2% HCl
CAS 7439-93-2, 7647-01-0, 7732-18-5
Density 0.53
Physical Form Solution
Molecular Formula Li
Concentration 995 to 1005 μg/mL of Li (1+) in 2% HCl (20°C)
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
SMILES [Li]
IUPAC Name lithium
Molecular Weight (g/mol) 6.94
Quantity 500 mL
PubChem CID 3028194
ChEBI CHEBI:30145
Formula Weight 6.94
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.