Learn More
Lithium sulfide, 99.9% (metals basis)
Description
It act as a superconductor and widely used in lithium-sulfur batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 900°C to 975°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 250 g |
| Assay Percent Range | 99.9% (Metal basis) |
| Solubility Information | soluble in water and ethanol |
| IUPAC Name | dilithium;sulfanide |
| Formula Weight | 45.95 |
| Odor | Like rotten eggs |
| Sensitivity | Moisture sensitive |
| Show More |
Frequently Asked Questions (FAQs)
Lithium sulfide releases flammable gas when in contact with water and should be kept away from water, moist air, or acids. Lithium sulfide should be stored and handled under a dry, inert gas atmosphere. Store at ambient temperatures, typically 20-25 degrees C.
Safety and Handling
| Hazard Category | H301-H314-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | DOT Class: 8; Packaging Group: II |
| EINECSNumber | 235-228-1 |
| RTECSNumber | OJ6439500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.