Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Tetraborate/Lithium Metaborate (35%/65%) Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™
SDP

Catalog No. FFB350002 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Triple-leached LDPE bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
FFB350002 1 kg Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FFB350002 Supplier SPEX CertiPrep Supplier No. FFB350002

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Lithium Tetraborate/Lithium Metaborate (35%/65%), Pure Grade Fusion Flux for Sample Preparation, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Pure Grade Fusion Flux
  • 1 kg
  • Lithium Tetraborate/Lithium Metaborate (35%/65%)

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Quantity 1 kg
Grade Pure
Concentration or Composition (by Analyte or Components) Various
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lithium Tetraborate/Lithium Metaborate, Fusion Flux
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.