Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tetraborate, 99.6% (metals basis)

Catalog No. p-4407589
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1259218 50 g
AA1259230 250 g
AA12592A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1259218 Supplier Thermo Scientific Chemicals Supplier No. 01259218
Only null left
Add to Cart

CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol

Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Melting Point 930°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.6% (Metal basis)
Solubility Information Moderately soluble in water.
Formula Weight 169.12
Appearance White powder
Density 2.4
Chemical Name or Material Lithium tetraborate
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 234-514-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.