Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Tetraborate, Solstice

Catalog No. 6002143 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-021-43 250 g
60-021-42 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-021-43 Supplier Solstice Supplier No. 222534250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

≥99.9% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Physical Form Powder
Quantity 250 g
Assay Percent Range 99.90%
Linear Formula Li2B4O7
UN Number NONH for all modes of transport
Synonym dilithium tetraborate
Formula Weight 169.12g/mol
Percent Purity ≥99.9% (trace metals basis)
Grade Reagent
Concentration or Composition (by Analyte or Components) 99.9% (trace metals basis)
Packaging Plastic Bottle
Product Line Solstice
Appearance White Powder
Density ≥0.25g/mL at 25°C
Assay Trace Metals Basis
Chemical Name or Material Lithium Tetraborate
Show More Show Less
EINECSNumber 234-514-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.