Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Tetraborate, Spectrum™ Chemical
SDP

Catalog No. 18611352 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-352 25 g Poly Bottle
18-611-354 125 g Poly Bottle
18-611-356 500 g Poly Bottle
18-611-355 2.5 kg Poly Pail
18-611-353 5 kg Poly Pail
18-611-357 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-352 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L113025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
  • Molecular Formula: Li2B4O7
  • Molecular Weight: 169.12 g/mol
  • CAS Number: 12007-60-2
  • Grade: Reagent
  • Molar Ratio (B2O3:Li2O): 1.95
  • Specifications: Iron (Fe) ≤ 0.03%, Sodium (Na) ≤ 0.015%, Calcium (Ca) ≤ 0.015%, Water ≤ 1.0%
  • Quality Standards: ISO 9001:2015 certified, manufactured under cGMP

Chemical Identifiers

CAS 12007-60-2

Specifications

Color White
CAS Max % 1
Physical Form Powder
Quantity 25 g
Molecular Formula Li2B4O7
Linear Formula Li2B4O7
UN Number NONH for all modes of transport
Synonym Dilithium tetraborate
IUPAC Name Dilithium tetraborate
Molecular Weight (g/mol) 169.12
ChEBI CHEBI:37335
Grade Reagent
Concentration or Composition (by Analyte or Components) Water: 1.0 %, Iron (Fe): 0.03 %, Sodium (Na): 0.015 %, Calcium (Ca): 0.015 %
Identification Passes Test
Packaging Poly Bottle
Appearance White Powder
Chemical Name or Material Lithium Tetraborate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.