Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Triethylborodeuteride, 1M Solution in THF, Thermo Scientific™

Catalog No. AC177701000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC177701000 100 mL
AC177705000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177701000 Supplier Thermo Scientific Chemicals Supplier No. 177701000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74540-86-6
Molecular Formula C6H16BLi
Molecular Weight (g/mol) 106.95
InChI Key WCJAYABJWDIZAJ-DRGWXPQLSA-N
Synonym lithium triethyl 2h hydroborate 1-, lithium triethylborodeuteride, borate 1-, triethylhydro-d-, lithium, t-4, super-deuteride® solution
PubChem CID 23664629
IUPAC Name lithium;deuterio(triethyl)boranuide
SMILES [Li+].[BH-](CC)(CC)CC

Specifications

Physical Form Liquid
Color Colorless
Quantity 100 mL
Density 0.94
Linear Formula LiB(C2H5)3D
Formula Weight 106.95
Chemical Name or Material Lithium triethylborodeuteride
Hazard Category HYGROSCOPIC
Hazard Statement MOISTURE SENSITIVE
EINECSNumber 277-915-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.