Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium trifluoromethanesulfonate, 98%

Catalog No. AC358710250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC358710250 25 g Glass bottle
AC358710050 5 g Glass bottle
AC358711000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC358710250 Supplier Thermo Scientific Chemicals Supplier No. 358710250
Only null left
Add to Cart

CAS: 33454-82-9 | CF3LiO3S | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33454-82-9
Molecular Formula CF3LiO3S
Molecular Weight (g/mol) 156.01
MDL Number MFCD00013227
InChI Key MCVFFRWZNYZUIJ-UHFFFAOYSA-M
Synonym lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
PubChem CID 3664839
IUPAC Name lithium;trifluoromethanesulfonate
SMILES [Li+].C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 423.0°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 4.34 to 4.56% (Li)
Packaging Glass bottle
Linear Formula F3CSO3Li
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 156.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Lithium trifluoromethanesulfonate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 251-528-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.