Learn More
NEOGEN Liver Digest, 500 g
SDP
Supplier:  NEOGEN 700004720
Encompass_Preferred
Liver Digest is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans.Liver digest is prepared by the controlled hydrolysis of liver. It is rich in vitamins and essential amino acids and has excellent nutritional properties and especially favors the growth of strict anaerobes and other fastidious microorganisms. Due to its capacity to stimulate sugar metabolism in saccharolytic organisms it is perfectly suited for the growth of a broad range of organisms such as Clostridium, Leuconostoc, Bacillus, homo and hetero-fermentative Lactic acid bacteria, as well as yeasts and filamentous molds.For more information visit https://www.neogen.com/solutions/Microbiology/liver-digest
Catalog No. 50-201-5858
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.