Learn More
Enzo Life Sciences Liver (human), (microsome extract) (200µg)
SDP
Supplier:  Enzo Life Sciences ADILYTHM100F
Encompass_Preferred
Formulation: Liquid. In SDS-PAGE sample buffer, pH 6.8, containing 40mM TRIS-HCl, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% bromophenol blue. Source: Pooled human liver samples. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. Applications: WB .
Catalog No. 50-200-7338
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.