Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Liver Tissue Slides (Diabetes)- Frozen
SDP

Catalog No. NBP259070 Shop All Novus Products
Change view
Click to view available options
Quantity:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259070 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259070 Supplier R&D Systems™ Supplier No. NBP259070

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Frozen tissue slides are ideal for rapidly identifying cellular localization of RNA or protein.

  • Species Human
  • Type: Tissue
  • Tissues: Liver
  • Tissue Condition: Diabetes

Specifications

Cross Reactivity Human
Type Liver Tissue Slides
Quantity 5
Regulatory Status RUO
Specimen Type Tissue
Tissue Condition Diabetes
Product Line Novus

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.