Learn More
Selleck Chemical LLC LJI308 5mg 1627709-94-7
SDP
Supplier:  Selleck Chemical LLC S78715MG
Encompass_Preferred
LJI308 is a potent, and pan-RSK (p90 ribosomal S6 kinase) inhibitor with IC50 of 6 nM, 4 nM, and 13 nM for RSK1, RSK2, and RSK3, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5360
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.