Learn More
Cayman Chemical LK6L9trIfluoroACTAte salt 10mg
SDP
Supplier:  Cayman Chemical 3884710 MG
Encompass_Preferred
A cytolytic peptide; cytotoxic to LNCaP-CL1, 22Rv1, and LNCaP cells (LC50s = 4, 4, and 6 µM, respectively), as well as non-cancerous NIH3T3 and OL cells (LC50s = 7 and 5 µM, respectively); induces hemolysis of isolated human erythrocytes at 100 µM
Catalog No. 50-291-8796