Learn More
CPC Scientific H-Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LL37003A
Encompass
LL-37 amide is an antimicrobial peptide with angiogenic activity.ONE-LETTER SEQUENCE: LLGDFFRKSKEKIGKEFKRIVQRIKDFLRNLVPRTES-NH2MOLECULAR FORMULA: C205H341N61O52MOLECULAR WEIGHT:4492.34STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [597562-32-8], RESEARCH AREA: AntimicrobialREFERENCES: S.Sandgren et al., J. Biol. Chem., 279, 17951 (2004); F.Neville et al., Biophys. J., 90, 1275 (2006); U.H.Dürr et al., Biochim. Biophys. Acta, 1758, 1408 (2006); T.Tecle et al., Innate Immun., 16, 151 (2010)
Catalog No. 50-210-9685