Learn More
Carbosynth LLC. LL-37 ( Human) (0.1mg vial), 1ea, LL-37 is a 37-amino acid peptide resulting from extracellular cleavage of the C-terminal end of the 18-kDa hCAP18 protein., MW: 4493.3, Molecular formula: C205H340N60O53, CAS# 154947-66-7
SDP
Supplier:  Carbosynth LLC. PLL4445S0.1MG
Encompass_Preferred
Synonyms: Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser, CAP-18, CAP18, LL37, LLGDFFRKSKEKIGKEFKRIVQRIKDFLRNLVPRTES, Application: Cathelicidin Antimicrobial Peptide, Storage Temperature: -20°C, References: G.H. Gudmundsson, B. Agerberth, J. Odeberg, T. Bergman, B. Olsson, and R. Salcedo,Eur. J. Biochem., 238, 325 (1996). (Original), R. Bals and J.M. Wilson, Cell. Mol. Life Sci., 60, 711 (2003). (Review), R. Lande, J. Gregorio, V. Facchinetti, B. Chatterjee, Y.-H. Wang, B. Homey, W. Cao, Y.-H. Wang, B. Su, F.O. Nestle, T. Zal, I. Mellman, J.-M. Schröder, Y.-J. Liu, and M. Gilliet, Nature, 449, 564 (2007). (Pharmacol.)
Catalog No. 50-168-6591