Learn More
Tocris Bioscience™ LL 37
GREENER_CHOICE
Catalog No. 52131
Change view
Click to view available options
1mg
Catalog No.
52-131 1mg
1 options
This item is not returnable. View return policy
Catalog No. 52-131 Supplier Tocris Bioscience™ Supplier No. 5213/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Antimicrobial peptide derivative of human cathelicidin
LL 37 is an antimicrobial peptide derivative of human cathelicidin. Induces FPRL1-mediated chemotaxis of human neutrophils, monocytes and T cells in vitro. Promotes wound healing following skin-targeted electroporation of a plasmid encoding hCAP-18/LL-37 in mice. LL 37 reduces SARS-CoV-2 infection by blocking the receptor binding domain of the S1 spike protein (Kd = 11.2 nM) and by binding to ACE2 (Kd = 25.5.nM). LL 37 inhibits SARS-CoV-2 pseudovirion infection (IC50 = 4.74 μg/mL) in vitro and in vivo. Also triggers apoptosis in colon cancer cells. Cell permeable.
Specifications
Molecular Weight (g/mol) 4493.32
Product Type LL 37
Quantity 1 mg