Learn More
Enzo Life Sciences LL-Z1640-2 (1mg). CAS: 66018-38-0
SDP
Supplier:  Enzo Life Sciences ALX380267M001
Encompass_Preferred
Inhibitor of the JNK/p38 pathways. Inhibits anisomycin-induced but not TNF-induced JNK/p38 activation. Potent protein kinase inhibitor.A naturally isolated cell-permeable fungal resorcylic lactone that acts as a selective and highly potent inhibitor against the MAPKKK TAK1 activity (IC50 = 8nM) in an ATP-competitive and irreversible manner; also inhibits MEK1 (IC50 = 411nM) and three other MAPKKKs (IC50 ≥268 nM against MEKK1, ASK1, and MEKK4) but at much higher concentrations. Shown to inhibit IL-1-induced/TAK1-mediated, but not H2O2-induced/TAK1-independent, JNK and p38 phosphorylation in 293 cells. Alternative name: (5Z)-7-Oxozeaenol. Purity: ≥95% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in methanol, acetone, DMSO (up to 25mM) or ethyl acetate; insoluble in ethanol and n-hexane. Long Term Storage: -20°C.
Catalog No. 50-200-8996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.