Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LLGL1 Polyclonal Antibody

Catalog No. PIPA554637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554637 Supplier Invitrogen™ Supplier No. PA554637

Rabbit Polyclonal Antibody

Immunogen sequence: FERFSLSARN ITEPLCSLDI NWPRDATQAS YRIRESPKLS QANGTPSILL APQSLDGSPD PAHSMGPDTP EPPEAALSPM SIDSATSADT TLDTTGDVTV EDVKDFLGSS EESEKNLRNL AEDE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 72%.

LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LLGL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LLGL1
Gene Accession No. Q15334
Gene Alias AI325176; DLG4; HUGL; Hugl protein; HUGL1; Hugl-1; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1 (Drosophila); lethal giant larvae homolog 1, scribble cell polarity complex component; lethal(2) giant larvae protein homolog 1; Lgl1; LLGL; Llgl1; LLGL1, scribble cell polarity complex component; Llglh; Mgl1; Mgl-1; Mlgl; Rgl1; Rgl-1
Gene Symbols LLGL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LLGL1. Recombinant protein control fragment (Product #RP-93255).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.