Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam LM11A-31, Nonpeptide p75NTR Ligand, 25 mg

Catalog No. 502704868
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4868 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4868 Supplier Abcam Supplier No. AB14618725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective nonpeptide p75NTR ligand that promotes neurogenesis and neurite survival.

  • Molecular Weight: 316.26 Da
  • Key Properties: Selective nonpeptide p75NTR ligand; blocks p75-mediated cell death; promotes neurogenesis and neurite survival
  • Binding Profile: Does not affect NGF binding to TrkA
  • Biological Activity: Neuroprotective and neuroregenerative effects in vivo; orally active; blood-brain barrier permeable
  • CAS Number: 1243259-19-9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Format Vial
Shipping Condition Ambient
Target Molecule p75NTR
Buffer 100% LM11A-31
Form Powder
Substrate Properties Chemical Substrate
Quantity 25 mg
For Use With (Application) Blocking p75-mediated cell death, neurogenesis, neurite survival
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.