Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Lm11a-31, Selective Nonpeptide P75NTR Ligand, 5 mg

Catalog No. 502704867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4867 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4867 Supplier Abcam Supplier No. AB1461875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Selective nonpeptide p75NTR ligand that promotes neurogenesis and neuroprotection with blood-brain barrier permeability.

  • Molecular Weight: 316.26 Da
  • Biological Activity: Selective nonpeptide p75NTR ligand; blocks p75-mediated cell death; promotes neurogenesis and neurite survival
  • Mechanism: Does not affect NGF binding to TrkA
  • Properties: Orally active, blood-brain barrier permeable; shows neuroprotective and neuroregenerative effects in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule p75NTR
Shipping Condition Ambient
Format Vial
Buffer 100% LM11A-31
Content And Storage Store at -20°C.
Quantity 5 mg
For Use With (Application) Neurogenesis, neurite survival, neuroprotection, neuroregeneration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.