Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101135 Supplier Invitrogen™ Supplier No. PA5101135
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LMO4.

LMO4 encodes a cysteine-rich protein that contains two LIM domains but lacks a DNA-binding homeodomain. The encoded protein may play a role as a transcriptional regulator or as an oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMO4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LMO4
Gene Accession No. P61968, P61969
Gene Alias A730077C12Rik; Breast tumor autoantigen; Crp3; ethanol induced 4; Etohi4; LIM domain only 4; LIM domain only protein 4; LIM domain transcription factor LMO4; LIM-only 4 protein; LMO4; LMO-4
Gene Symbols LMO4
Host Species Rabbit
Immunogen A synthesized peptide derived from human LMO4(Accession P61968), corresponding to amino acid residues C118-C165.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16911, 362051, 8543
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.