Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMTK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114265 Supplier Invitrogen™ Supplier No. PA5114265
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGLLKSPRAA DEPEDSELER KRKMVSFHGD VTVYLFDQET PTNELSVQGT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%.

Protein kinase which phosphorylates ESR1 (in vitro) and protects it against proteasomal degradation. May also regulate ESR1 levels indirectly via a PKC-AKT-FOXO3 pathway where it decreases the activity of PKC and the phosphorylation of AKT, thereby increasing binding of transcriptional activator FOXO3 to the ESR1 promoter and increasing ESR1 transcription (PubMed:21602804). Involved in endocytic trafficking of N-methyl-D-aspartate receptors (NMDAR) in neurons. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMTK3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LMTK3
Gene Accession No. Q5XJV6
Gene Alias Aatyk3; Apoptosis-associated tyrosine kinase 3; BC059845; KIAA1883; Lemur tyrosine kinase 3; LMR3; Lmtk3; LOC691056; PPP1R101; protein phosphatase 1, regulatory subunit 101; serine/threonine-protein kinase LMTK3; similar to 5E5 antigen; TYKLM3
Gene Symbols LMTK3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of Mouse Lmtk3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381983
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.