Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMX1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115517 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115517 Supplier Invitrogen™ Supplier No. PA5115517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LMX1A.

The LIM homeobox transcription factor 1 (LMX1A) belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1A binds an A/T-rich sequence and functions in the nucleus as a transcriptional activator to the insulin gene promoter. In the developing embryo, LMX1A is expressed along the neuraxis and leads to the development of the roof plate of the vertebrae. While LMX1A is crucial for the differentiation of dopamine neurons, the related LMX1B appears to be important for both the development and the survival of these cells. It has been suggested that LMX1A might be a useful biomarker or risk factor in tumor aggressiveness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMX1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Lmx1a
Gene Accession No. Q8TE12, Q9JKU8
Gene Alias dr; dreher; LIM homeobox transcription factor 1 alpha; LIM homeobox transcription factor 1, alpha; LIM homeobox transcription factor 1-alpha; LIM/homeobox protein 1.1; LIM/homeobox protein LMX1A; LMX1; LMX-1; Lmx1.1; LMX-1.1; Lmx1a; MGC87616; shaker short-tail; sst
Gene Symbols Lmx1a
Host Species Rabbit
Immunogen A synthesized peptide derived from human LMX1A(Accession Q8TE12), corresponding to amino acid residues R152-L202.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110648, 289201, 4009
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.