Learn More
Cayman Chemical Lnnamvr 25mg
SDP
Supplier:  Cayman Chemical 3913925 MG
Encompass_Preferred
An FHIT inhibitor (IC50 0.12 UM) inhibits the growth of HCC827 and H460 cells (GI50s 0.87 and 5.9 UM respectively)
Catalog No. 50-291-8395
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.