Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LNX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PI720003 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720003 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720003 Supplier Invitrogen™ Supplier No. 720003
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Rat, Mouse and Goat.

Lnx1 (Ligand of Numb Protein X 1) protein is encoded by the Lnx1 gene located on chromosome 4 in humans and is known as PDZRN2 (PDZ domain-containing RING finger 2). It is through its RING finger and PDZ domains that Lnx1 forms oligomers and interacts with some of its target proteins such as JAM4 to form multiprotein complexes. Lnx1 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Phosphotyrosine binding domain (PTB) on Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus positively regulates the Notch signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LNX1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2-7.4
Gene LNX1
Gene Accession No. O70263, Q8TBB1
Gene Alias E3 ubiquitin-protein ligase LNX; Ligand of Numb protein X 1; ligand of Numb-binding protein 1; Ligand of Numb-protein X 1; ligand of numb-protein X 1, E3 ubiquitin protein ligase; LNX; Lnx1; MPDZ; multi-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX; numb-binding protein 1; PDZ domain-containing RING finger protein 2; PDZRN2; RING-type E3 ubiquitin transferase LNX; UNQ574/PRO1136
Gene Symbols LNX1
Host Species Rabbit
Immunogen Multiple peptides against Human LNX1 (aa 200-213, 704-719).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16924, 360926, 84708
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.