Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Loading Control (Simple Western) Antibody Pack, Novus Biologicals™
SDP

Catalog No. NB183504 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
4 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183504 4 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183504 Supplier R&D Systems Supplier No. NBP306859

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Loading Control (Simple Western) Monoclonal specifically detects Loading Control (Simple Western) in Human/Mouse/Rat samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Simple Western/Western Blot.

Specifications

Antigen Loading Control (Simple Western)
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Simple Western, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Gene Alias GAPDH, Hsp60
Host Species Mouse
Quantity 4 Vials
Regulatory Status RUO
Research Discipline Loading Controls
Target Species Human, Mouse, Rat
Content And Storage Store at 4°CC.
Product Type Antibody Packs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.