Learn More
Blackline Safety™ Location Beacon, North America
Catalog No. 170001975 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Type:
Location Beacon
Catalog No. Type
17-000-1975 Location Beacon
1 options
Catalog No. 17-000-1975 Supplier Blackline Safety™ Supplier No. BEACONNA
  • $311.00
    $218.65 / Each of 1
    Save $92.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Provides an additional positioning signal to accurately map the locations of employees and alerts.
  • Compact, wireless, self-powered positioning signal transmitters
  • Ideal for use in areas where GPS may be inconsistent or unavailable
  • Beacons broadcasts a unique ID number that is received by nearby G7 devices
  • Beacon locations can be recorded on custom floor or site plans in Blackline Live, providing valuable situational context
  • Precise positioning empowers the fastest emergency response — time is never wasted looking in the wrong places
Specifications
Type Location Beacon