Learn More
Brady™ Lockable Ratchet Straps for Tower Skirt Confined Space Cover
Shop All Brady Corporation ProductsDescription
- Sturdy straps with lockable option enhance usability
- Compatible with Confined Space Safety Cover Tower Skirt Kits to clearly signal an unavoidable boundary
Specifications
Specifications
| Product Type | Lockable Ratchet Straps |
| Color | Black |
| Material | Nylon |
| Description | Wraps up to 8 ft. diameter |
| For Use With (Equipment) | Confined Space Cover |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
