Learn More
Post Medical Locking-Top Sharps Containers
Catalog No. PM1101LPBW6
Change view
Click to view available options
Capacity (English):
1.5 qt.
2 qt.
1.5 gal.
2.5 gal.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
PM1101LPBW6 2.5 gal.
PMSL200 2 qt.
PM1101LPR 2.5 gal.
PMSL150 1.5 qt.
PM2201LPR 1.5 gal.
PM2201LPBW8 1.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. PM1101LPBW6 Supplier Post Medical Supplier No. 1101LPBW16
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safe, one-handed sharps disposal
For use in patient rooms and all hospital departments including laboratory.
  • Thick-walled, puncture-resistant sharps containment systems provide safe, convenient and affordable site of origin sharps disposal
  • Vertical drop disposal minimizes excessive handling; reduces risks associated with horizontal “mail-box” type disposal
  • Once self-contained screw cap is secured and locked, leakage of fluids during handling, transport and disposal is virtually eliminated
  • 1.0, 1.5 and 2.5 gal. Containers
  • Can be used freestanding or with metal stands or cabinets
  • Bulk 3.5 and 6 gal. Containers
  • Designed for freestanding use only
Compliance
Meets OSHA requirements.
Specifications
Capacity (Metric) 9.5 L
Closure Type Locking Screw Cap
Capacity (English) 2.5 gal.
For Use With (Application) For use in patient rooms and all hospital departments including laboratory.