Learn More
Post Medical Locking-Top Sharps Containers
Description
For use in patient rooms and all hospital departments including laboratory.
- Thick-walled, puncture-resistant sharps containment systems provide safe, convenient and affordable site of origin sharps disposal
- Vertical drop disposal minimizes excessive handling; reduces risks associated with horizontal “mail-box” type disposal
- Once self-contained screw cap is secured and locked, leakage of fluids during handling, transport and disposal is virtually eliminated
- 1.0, 1.5 and 2.5 gal. Containers
- Can be used freestanding or with metal stands or cabinets
- Bulk 3.5 and 6 gal. Containers
- Designed for freestanding use only
Compliance
Meets OSHA requirements.
Specifications
Specifications
| Capacity (Metric) | 5.7 L |
| Closure Type | Locking Screw Cap |
| Capacity (English) | 1.5 gal. |
| For Use With (Application) | For use in patient rooms and all hospital departments including laboratory. |