Learn More
Brady™ Lockout Tagout Station, Cabinet Only with Locking Handle
Catalog No. 19103990 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Yellow
Catalog No. Color
19-103-990 Black on Yellow
1 options
Catalog No. 19-103-990 Supplier Brady™ Supplier No. LC977A
Add to Cart
Add to Cart
Lockout Stations are a great way to centralize lockout activity needs by organizing the tools, padlocks and padlock key in one location. The Brady™ Lockout Tagout Station, Cabinet Only with Locking Handle has a clear plastic window that allows for an easy viewing of contents.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Product Type Cabinet with Carrying Handle
Color Black on Yellow
Material Powder-coated Steel
Includes Opaque door
Quantity 1 Ea.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.