Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Lockout Tagout Tags: DO NOT OPERATE I certify that the following tasks have been performed to lockout this equipment REV A

Catalog No. 19113274 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Economy Polyester (B-851)
Heavy-duty Polyester
Paper Cardstock
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-113-274 Economy Polyester (B-851)
19-113-271 Heavy-duty Polyester
19-113-260 Paper Cardstock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-113-274 Supplier Brady™ Supplier No. 66052
Add to Cart
Add to Cart

Write-On Paper Cardstock: Surface printed Cardstock (B-853) tags provide an economical, yet durable material option for temporary tagging in a non harsh environment.

  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Withstands moisture, grease and dirt
  • Stays legible longer than cardstock

Specifications

Product Type Tag
Color Black/Red on White
Material Economy Polyester (B-851)
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147
Dimensions (L x W) 5.75 x 3 in.
Height (English) 5.75 in.
Quantity 25 Pk.
Width (English) 3.6 in.
Width (Metric) 91.44 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.