Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Lockout Tagout Tags: THIS TAG & LOCK OUT REMOVED ONLY BY PERSON SHOWN ON BACK ESTA ETIQUETA Y SEGUIRO

Catalog No. 19113234 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
THIS TAG and LOCK OUT REMOVED ONLY BY PERSON SHOWN ON BACK ESTA ETIQUETA Y SEGURO
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-113-234 THIS TAG and LOCK OUT REMOVED ONLY BY PERSON SHOWN ON BACK ESTA ETIQUETA Y SEGURO
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-113-234 Supplier Brady™ Supplier No. 48797
Add to Cart
Add to Cart

Economy Polyester (B-851) tags are perfect for marking on-the-spot warnings of dangerous conditions.

  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Withstands moisture, grease and dirt
  • Stays legible longer than cardstock

Specifications

Product Type Tag
Color Black/Red on White
Material Polyester
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147
Dimensions (L x W) 5.75 x 3 in.
Displayed Text THIS TAG and LOCK OUT REMOVED ONLY BY PERSON SHOWN ON BACK ESTA ETIQUETA Y SEGURO
Height (English) 5.75 in.
Includes (25) Nylon Fasteners
Quantity 25 Pk.
Width (English) 3.6 in.
Width (Metric) 91.44 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.