Learn More
Vernier™ Logger Pro 3 (Download)
SDPSCI_ED
Catalog No. S16184ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Educational use only
Catalog No. For Use With (Application)
S16184ND Educational use only
1 options
Catalog No. S16184ND Supplier Vernier™ Supplier No. LPE
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Logger Pro 3 (Download)
Data-collection and analysis software for Windows and Mac computers.
  • 12 built-in, self-guided tutorials.
  • Write labs in Logger Pro or export data to Word or Excel.
  • Includes over 1,000 experiment files supported by our library of lab books.
  • Develop your own experiment files to match your curriculum.
  • Supports over 80 Vernier sensors for real-time graphing and analysis.
  • Collect data from a variety of sources, including LabQuest 2, LabQuest Stream, LabQuest Mini, LabPro, WDSS, spectrometers, Ohaus balances, ProScope HR, and more.
  • Enter values manually from existing data tables or import data.
  • Export to Plotly and LabArchives.
  • Capture videos to add a visual record of an experiment synchronized with your data.
  • Analyze a video frame by frame or take measurements from a still photo.
  • Draw predictions on graphs prior to data collection.
  • Perform statistical analysis of data, including integrals, tangents, curve fits (pre-defined, custom, and weighted), and more.
  • Auto-ID sensors make setup effortless.
  • Collect live data from more than 80 different sensors and devices.
  • Draw predictions on a graph before collecting data.
  • Use a variety of data-collection modes, as needed, for your experiment: time-based data, selected events, events with typed-in entries, photogate, radiation counting, and more.
  • Manually enter data for graphing and analysis.
  • Import data from Vernier interfaces.
  • Lay out graphs, tables, and text as needed across multiple pages to describe your experiment.
  • Read values and slope from graphs using examine and tangent line tools.
  • Print graphs and data tables.
  • XY graphs, log graphs, double-Y graphs, strip charts, and FFT graphs
  • Model data with user-adjustable functions.
  • Extract data from movies using frame-by-frame video analysis.
  • Capture videos from DV cameras and web cameras.
  • Play back movies synced to sensor data.
  • Gel electrophoresis analysis
  • Calculated columns allow you to graph new quantities such as kinetic energy.
  • Time of day and date stamps for graphing
  • Ocean Optics spectrometer support
  • GPS support
  • GC peak analysis
  • Manual configuration mode for IB courses
Warranty and Services
5-year limited warranty
Specifications
Type Data Collection Software
Includes Software CD, Site License, free software updates, quick reference manual
For Use With (Application) Educational use only