Learn More
Labconco Logic A2 Airflow Sensor
SDP
Catalog No. 13118377 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8 ft. Logic Biosafety Cabinet
Catalog No. For Use With (Equipment)
13-118-377 8 ft. Logic Biosafety Cabinet
1 options
Catalog No. 13-118-377 Supplier Labconco Supplier No. 3277618
Add to Cart
Add to Cart
Airflow sensor kit for recirculating 8 ft. Logic Biosafety Cabinets.
  • Airflow sensors display both inflow and downflow velocities on the line-of-sight touchscreen display.
  • Includes two sensors for an exhausted 8 ft. Logic Biosafety Cabinet.
  • Certifier installation and calibration required.
Specifications
Product Type Airflow Sensor
For Use With (Equipment) 8 ft. Logic Biosafety Cabinet