Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Logic™ Cell Culture Cart Pipette Insert Accessory
SDP

Catalog No. 01812404 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Logic Cell Culture Cart
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-812-404 Logic Cell Culture Cart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-812-404 Supplier Labconco™ Supplier No. 8024600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Divides the lower storage area of Logic Cell Culture Cart into six compartments for storing various size serological pipettes.

  • Insert adds six compartments to lower storage are of the Logic Cell Culture Cart.
  • Ideal for additional storage of serological pipettes.

Specifications

Description Pipette Insert accessory adds 6 extra compartments for storing various size serological pipettes in the Logic Cell Culture Cart.
Width (Metric) Interior 39.4 cm
Product Type Pipette Insert
Width (English) Exterior 15.5 in.
Width (Metric) Exterior 39.4 cm
Includes Pipette Insert, Installation Instructions
Width (English) Interior 15.5 in.
For Use With (Equipment) Logic Cell Culture Cart
Depth (English) 15.5 in.
Depth (Metric) 39.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.