Learn More
Labconco Logic IV Bar Kit
SDP
Catalog No. 13118378 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8 ft. Logic Biosafety Cabinet
Catalog No. For Use With (Equipment)
13-118-378 8 ft. Logic Biosafety Cabinet
1 options
Catalog No. 13-118-378 Supplier Labconco Supplier No. 3288904
Add to Cart
Add to Cart
IV bar kit for 8 ft. Logic Biosafety Cabinets.
  • IV bar kit for an 8 ft. Logic Biosafety Cabinet.
  • Includes IV bar, mounting hardware, and eight IV hooks: four short and four long.
  • Constructed of stainless steel.
Specifications
Product Type IV Bag Holder Kit
For Use With (Equipment) 8 ft. Logic Biosafety Cabinet