Learn More
Labconco™ Logic™ Mag-Zone Magnetic Accessories
SDP
Catalog No. 01812563 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Magnetic Pipette Aid Holder
Catalog No. Includes
01-812-563 Magnetic Pipette Aid Holder
1 options
Catalog No. 01-812-563 Supplier Labconco™ Supplier No. 3033200
Add to Cart
Add to Cart
Mag-Zone accessories attach to the right side wall of Logic Biosafety Cabinets and provide easy, cleanable organization for common tools used in biosafety cabinets.
  • Grade 304, polished stainless steel for easy disinfection. Strong magnets ensure secure attachment to the right side wall of Optic White Logic and PuriCare biosafety cabinets.
  • Three Accessories Available:
  • Pipette Rack - Holds 4 standard pipettors and one pipette aid
  • Pipette Aid Holder
  • Waste Bag Holder
Specifications
For Use With Pipettes
Type Pipette Holder
For Use With (Application) Hold/Store Pipette Aid in biosafety cabinet
For Use With (Equipment) Optic White Labconco Logic Biosafety Cabinets
Includes Magnetic Pipette Aid Holder
Dimensions (L x W x H) 8.6 x 5.3 x 7.0 cm