Learn More
Labconco™ Logic Vue Computer Arm
SDP
Catalog No. 10304209 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Logic Vue
Catalog No. For Use With (Equipment)
10-304-209 Logic Vue
1 options
This item is not returnable. View return policy
Catalog No. 10-304-209 Supplier Labconco™ Supplier No. 3339000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Computer arm that attaches directly to left side of Logic Vue and supports monitor, keyboard and mouse. Space-saving design eliminates need for separate computer cart.
  • Attaches directly to left side of Logic Vue and supports monitor, keyboard and mouse.
  • Space-saving design eliminates need for separate computer cart.
  • VESA design supports monitors up to 18 lb.
  • Laptop Computer Arm securely supports a laptop computer within easy view of an operator.
  • Articulating arm raises, lowers and pivots to a steady position.
  • Includes articulating arm and supportive laptop platform with mounting hardware.
  • Installation is required
Specifications
Product Type Computer Arm
For Use With (Equipment) Logic Vue
WARNING: Cancer - www.P65Warnings.ca.gov