Learn More
Labconco™ Logic Vue Connection Kit
SDP
Catalog No. 10304210 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Logic Vue Class II Enclosure
Catalog No. For Use With (Equipment)
10-304-210 Logic Vue Class II Enclosure
1 options
This item is not returnable. View return policy
Catalog No. 10-304-210 Supplier Labconco™ Supplier No. 3037000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Kit to connect two Logic Vue Class II Enclosures together for an expanded work area.
  • Connects two Logic Vue Class II Enclosures together for an expanded work area.
  • Connection provides a passthrough that is 38.9 x 28 in. (H x D) and 10 in. (W).
  • Installation is required.
  • Accessories for use with Logic Vue Class II Enclosures.
Specifications
Product Type Connection Kit
For Use With (Equipment) Logic Vue Class II Enclosure
WARNING: Cancer - www.P65Warnings.ca.gov