Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lomitapide 98.0+%, TCI America™
SDP

Catalog No. L0298100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0298100MG 100 mg
L029825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0298100MG Supplier TCI America Supplier No. L0298100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 182431-12-5
Molecular Formula C39H37F6N3O2
Molecular Weight (g/mol) 693.734
MDL Number MFCD16620494
InChI Key MBBCVAKAJPKAKM-UHFFFAOYSA-N
Synonym N-(2,2,2-Trifluoroethyl)-9-[4-[4-[4′-(trifluoromethyl)-[1,1′-biphenyl]-2-ylcarboxamido]piperidin-1-yl]butyl]-9H-fluorene-9-carboxamide, AEGR-733, BMS-201038
PubChem CID 9853053
ChEBI CHEBI:72297
IUPAC Name N-(2,2,2-trifluoroethyl)-9-[4-[4-[[2-[4-(trifluoromethyl)phenyl]benzoyl]amino]piperidin-1-yl]butyl]fluorene-9-carboxamide
SMILES C1CN(CCC1NC(=O)C2=CC=CC=C2C3=CC=C(C=C3)C(F)(F)F)CCCCC4(C5=CC=CC=C5C6=CC=CC=C64)C(=O)NCC(F)(F)F

Specifications

Melting Point 142°C
Color White
Quantity 100 mg
Formula Weight 693.73
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Lomitapide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.