Learn More
MilliporeSigma™ Supelco™ Long Stem Flow Control Valves
Catalog No. 111004107 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
6/Pk.
Catalog No. Packaging Quantity
11-100-4107 6/Pk.
1 options
Catalog No. 11-100-4107 Supplier MilliporeSigma™ Supelco™ Supplier No. 57048
Add to Cart
Add to Cart
Equip alternative valves in your 12-port or 24-port standard Visiprep vacuum manifold with these long stem flow control valves if you intend to use all ports of the manifold with 12, 20, or 60 mL SPE tubes. Note: Not for use with Visiprep DL (Disposable Liner) models
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 6/Pk.