Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LONP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552618 Supplier Invitrogen™ Supplier No. PA552618
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TILGVIPNTP DPASDAQDLP PLHRIGTAAL AVQVVGSNWP KPHYTLLITG LCRFQIVQVL KEKPYPIAEV EQLDRLEEFP NTCKMREELG ELSEQFYKYA VQLVEMLDMS VPAVAKLRR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

ATP-dependent serine protease that mediates the selective degradation of misfolded and unassembled polypeptides in the peroxisomal matrix. Necessary for type 2 peroxisome targeting signal (PTS2)-containing protein processing and facilitates peroxisome matrix protein import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LONP2
Gene Accession No. Q86WA8
Gene Alias 1300002A08Rik; AU015403; HAMAP-Rule:MF_03121}; Lon peptidase 2; lon peptidase 2, peroxisomal; lon protease 2; lon protease 2 {ECO:0000255; Lon protease homolog 2, peroxisomal; lon protease homolog 2, peroxisomal {ECO:0000255; lon protease-like protein 2; lon protease-like protein 2 {ECO:0000255; LONP; Lonp2; LONPL; LRRGT00016; MGC4840; Peroxisomal Lon protease; peroxisomal Lon protease {ECO:0000255; peroxisomal Lon protease homolog 2; peroxisomal LON protease like; Peroxisomal Lon protease-like protein 2-like protein; pLon; RGD1305466
Gene Symbols LONP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LONP2. Recombinant protein control fragment (Product #RP-89241).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.