Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ AccuGENE™ 0.5 M EDTA Solution

Catalog No. BMA51201 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA51201 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA51201 Supplier Lonza™ Supplier No. 51201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AccuGENE Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For R and D/experimental use only, wide range of molecular biology applications/techniques
pH 7 to 9
Quantity 100mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.