Learn More
Lonza™ AccuGENE™ 1M Tris HCl Buffers
Description
- Prepared with 18 megohm water
- Filtered using 0.2μ filter and filled into sterile bottles
- Manufactured according to strict quality control standards to ensure consistency
- Guaranteed DNase, RNase and protease-free
- Customized solutions are available to meet individual needs
Specifications
Specifications
| Concentration | 1 M |
| DNase | Yes |
| For Use With (Application) | Ideal for a wide range of molecular biology applications |
| pH | 7.2 |
| Quantity | 1000 mL |
For research or manufacturing use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.