Learn More
AccuGENE™ TAE Buffers, Lonza™
Description
- Ready-to-use
- Prepared with high quality reagents and use 18megohm water
- Filtered using 0.2μ filter
- Available in 10X and 50X strength
- Manufactured according to strict quality control standards to ensure consistency
- Customized solutions are available to meet individual needs
Specifications
Specifications
| Chemical Name or Material | DNA Sample Loading Buffer |
| Concentration | 10X |
| Physical Form | Liquid |
| Quantity | 4 L |
| Formulation | TAE Buffer, 10X with 0.4M Tris acetate, 0.01M EDTA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.