Learn More
Lonza™ DNA Ladders
Specifications
Specifications
| Concentration | 200 ng/μL |
| Form | Liquid |
| Formulation | Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA |
| For Use With (Application) | Size determination of DNA samples |
| Increments | 500bp |
| No. of Bands | 16 |
| Quantity | 300 μL |
| Storage Requirements | 2° to 8°C |
| Product Type | DNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.