Learn More
Lonza™ FlashGel™ DNA Starter Kit
Catalog No. BMA57063 Shop All Lonza, Inc Products
Change view
Click to view available options
Includes:
FlashGel DNA Cassettes, 1.2% agarose (9 Pk.); 12 + 1 well single tier; FlashGel Loading Dye, and FlashGel Marker (100 – 4kbp)
Catalog No. Includes
BMA57063 FlashGel DNA Cassettes, 1.2% agarose (9 Pk.); 12 + 1 well single tier; FlashGel Loading Dye, and FlashGel Marker (100 – 4kbp)
1 options
This item is not returnable. View return policy
Catalog No. BMA57063 Supplier Lonza™ Supplier No. 57063
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
FlashGel DNA Cassettes, 1.2% agarose (9 Pk.); 12 + 1 well single tier; FlashGel Loading Dye, and FlashGel Marker (100 – 4kb)
Load samples, watch bands migrate and get data in as little as two minutes
  • Provide nearly instant results
  • Needs no gel preparation, band excision, purification, and UV light
  • Complete separation, recovery and documentation safely at the bench
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Includes FlashGel DNA Cassettes, 1.2% agarose (9 Pk.); 12 + 1 well single tier; FlashGel Loading Dye, and FlashGel Marker (100 – 4kbp)