Learn More
Lonza™ FlashGel™ System Components and Packs: FlashGel Power Supply Pack
Catalog No. BMA57069 Shop All Lonza, Inc Products
Change view
Click to view available options
Includes:
FlashGel Dock and FlashGel Power Supply
Catalog No. Includes
BMA57069 FlashGel Dock and FlashGel Power Supply
1 options
Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.
Catalog No. BMA57069 Supplier Lonza™ Supplier No. 57069
Add to Cart
Add to Cart
Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.
Fast analysis, recovery, and documentation of DNA and RNA
  • Provides nearly instant results
  • Load samples, watch bands migrate and get data in as little as two minutes
  • Includes DNA recovery and gel documentation
  • Needs no gel preparation, band excision, purification, and UV light
  • Complete separation, recovery and documentation safely at the bench
  • Highly sensitive system allows a 5 to 20X reduction in DNA levels, saving time and money
  • Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
  • Detect <0.1ng DNA or <10ng total RNA
  • Clean, sharp separation and straight, uniform sample lanes
FlashGel Power Supply Pack
  • Includes: FlashGel Dock and FlashGel Power Supply
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Type FlashGel Power Supply Pack
Includes FlashGel Dock and FlashGel Power Supply