Learn More
Lonza™ FlashGel™ System Kit
Catalog No. BMA57067 Shop All Lonza, Inc Products
Change view
Click to view available options
Includes:
FlashGel Dock, a FlashGel Camera, a 9 Pk. of 1.2% DNA Cassettes, Loading Dye and Marker
Catalog No. Includes
BMA57067 FlashGel Dock, a FlashGel Camera, a 9 Pk. of 1.2% DNA Cassettes, Loading Dye and Marker
1 options
Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.
Catalog No. BMA57067 Supplier Lonza™ Supplier No. 57067
Add to Cart
Add to Cart
Request a Quote
Caution should be exercised in the operation of this system as it can develop sufficient voltage and current to produce a lethal shock. To avoid risk of injury, the system should only be operated by properly trained personnel and always in accordance with instructions provided with the FlashGel Dock.
Fast analysis, recovery, and documentation of DNA and RNA
  • Provides nearly instant results
  • Load samples, watch bands migrate and get data in as little as two minutes
  • Includes DNA recovery and gel documentation
  • Needs no gel preparation, band excision, purification, and UV light
  • Complete separation, recovery and documentation safely at the bench
  • Highly sensitive system allows a 5 to 20X reduction in DNA levels, saving time and money
  • Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
  • Detect <0.1ng DNA or <10ng total RNA
  • Clean, sharp separation and straight, uniform sample lanes
FlashGel System
  • Consists of enclosed, disposable, precast agarose gel cassettes and a combination unit
  • Includes: FlashGel Dock; FlashGel Camera; 9 Pk. FlashGel DNA Cassettes (1.2%, 12 + 1 well); FlashGel Loading Dye and FlashGel DNA Marker
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Type FlashGel System
Includes FlashGel Dock, a FlashGel Camera, a 9 Pk. of 1.2% DNA Cassettes, Loading Dye and Marker